Order Entry
Export
ContactUsLinkComponent
Sulphur 99.5%, resublimed
Irritant
Sulphur 99.5%, resublimed
Certificates
undefined
Irritant
Sulphur 99.5%, resublimed

Specification Test Results

  • Appearance:
    Yellow Powder
  • Assay:
    ≥99.4 %
  • Sulfated ash:
    ≤0.2 %
  • Free acid (H2SO4):
    ≤0.02 %
  • Free alkali (NaOH):
    ≤0.01 %

Specification Test Results

Related Information