
Sulphur 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7704-34-9
:
Formula:S
MDL Number:MFCD00085316
:32.07 g/mol
Merck Index:13,09059
Boiling Pt:445 °C (1013 hPa)
Melting Pt:114 °C
Density:2,36 g/cm³ (20 °C)
Flash Pt:168 °C
Storage Temperature:Ambient
EINECS:231-722-6
UN:1350
ADR:4.1,III
:GHS07
Specification Test Results
- Appearance:Yellow Pieces
- Particle size:≤12 mm
- Total trace metal impurities:≤0.001 %