Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bismuth
Bismuth, Sigma-Aldrich®
Bismuth, Sigma-Aldrich®
Bismuth, Sigma-Aldrich®
SIAL556130-250G
:
Merck
:
Bismuth, Sigma-Aldrich®
SIAL556130-250G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
250 g
Seq:
3545
:
:
208.98 g/mol
:
12,01297
:
1490 °C (1013 hPa)
:
271…272 °C
:
9.78 g/cm³ (20 °C)
:
231-177-4
:
7440-69-9
:
Bi
:
MFCD00134033