Bismuth, Sigma-Aldrich®
SIAL452386-50G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
:
:208.98 g/mol
:12,01297
:1490 °C (1013 hPa)
:271…272 °C
:9.78 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:231-177-4
:7440-69-9
:Bi
:MFCD00134033