Bismuth, Sigma-Aldrich®
Catalog # SIAL265470-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:208.98 g/mol
Merck Index:12,01297
Boiling Pt:1490 °C (1013 hPa)
Melting Pt:271…272 °C
Density:9.78 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:231-177-4
CAS Number:7440-69-9
Formula:Bi
MDL Number:MFCD00134033