

Antimony 1-5 mm, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-36-0
:
Formula:Sb
MDL Number:MFCD00134030
:121.76 g/mol
Merck Index:12,00733
Boiling Pt:1637 °C (1013 hPa)
Melting Pt:630 °C
Density:6.684 g/cm³ (20 °C)
UN:2871
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:231-146-5
:GHS07
:GHS09