Silicon
Catalog # PROONIST-57B
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:40 g
- Seq:3344
Specifications
About this item
:
:28.09 g/mol
Merck Index:12,08635
Boiling Pt:2355 °C (1013 hPa)
Melting Pt:1410 °C
Density:2.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-130-8
CAS Number:7440-21-3
Formula:Si
MDL Number:MFCD00085311