Silicon ≥99.999% (metals basis), plate, Thickness: 12.5 mm (0.5 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-21-3
:
Formula:Si
MDL Number:MFCD00085311
:28.09 g/mol
Merck Index:12,08635
Boiling Pt:2355 °C (1013 hPa)
Melting Pt:1410 °C
Density:2.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-130-8