Graphite, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:7782-42-5
- Formula:C
- MDL Number:MFCD00144065
- Molecular Weight:12.01 g/mol
- Merck Index:12,04560
- Melting Pt:3827 °C
- Density:2.09…2.23 g/cm³ (20 °C)
- EINECS:231-955-3
2 Options Available Below
Product Details & Documents
- CAS Number:7782-42-5
- Formula:C
- MDL Number:MFCD00144065
- Molecular Weight:12.01 g/mol
- Merck Index:12,04560
- Melting Pt:3827 °C
- Density:2.09…2.23 g/cm³ (20 °C)
- EINECS:231-955-3



