Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glassy carbon
Glassy carbon, rods, Ø 6 mm (0.24 in) Type 1
Glassy carbon, rods, Ø 6 mm (0.24 in) Type 1
:
Thermo Fisher Scientific
Glassy carbon, rods, Ø 6 mm (0.24 in) Type 1
:
Thermo Fisher Scientific
:
7440-44-0
:
:
C
:
MFCD00133992
:
12.01 g/mol
:
13,01818
:
3652 °C
:
1.95 g/cm³ (20 °C)
:
Ambient
:
231-153-3
msds_ALFA045004