Order Entry
Export
Orders LinkContactUsLinkComponent
Charcoal activated

Charcoal activated

Supplier:   Roth Carl
Charcoal activated
Supplier:  Roth Carl
Certificates
Charcoal activated

3 Options Available Below

Product Details & Documents
Synonyms:  Carbon activated, Activated carbon, Activated charcoal
  • CAS Number:7440-44-0
  • Formula:C
  • MDL Number:MFCD00133992
  • Molecular Weight:12.01 g/mol
  • Merck Index:13,01818
  • Melting Pt:3550 °C
  • Density:2 g/cm³ (20 °C)
  • EINECS:231-153-3
  • UN:1362
  • ADR:4.2,III
  • REACH:01-2119488894-16
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 69,50 €
    69,50 €
    1 * 1 kg
       69,50 €

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 244,00 €
    244,00 €
    1 * 5 kg
       244,00 €