Order Entry
Export
ContactUsLinkComponent
Indium
Indium
Catalog # SIAL57083-10G-F
Supplier:  Merck
CAS Number:  
Indium
Catalog # SIAL57083-10G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :114.82 g/mol
Merck Index:13,04971
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:156 °C
Density:7.3 g/cm³ (20 °C)
EINECS:231-180-0
CAS Number:7440-74-6
Formula:In
MDL Number:MFCD00134048