Indium
Catalog # ROTH9803.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:114.82 g/mol
Merck Index:13,04971
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:156 °C
Density:7.3 g/cm³ (20 °C)
EINECS:231-180-0
CAS Number:7440-74-6
Formula:In
MDL Number:MFCD00134048