Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium
Magnesium ≥99.5% purum, turnings for grignard-reaction, Sigma-Aldrich®
Magnesium ≥99.5% purum, turnings for grignard-reaction, Sigma-Aldrich®
Magnesium ≥99.5% purum, turnings for grignard-reaction, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Magnesium ≥99.5% purum, turnings for grignard-reaction, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7439-95-4
:
Formula:
Mg
MDL Number:
MFCD00085308
:
24.3 g/mol
EINECS:
231-104-6
:
GHS02
Storage Temperature:
Ambient