Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium
Magnesium ≥99.8% (metals basis), turnings
Magnesium ≥99.8% (metals basis), turnings
:
Thermo Fisher Scientific
Magnesium ≥99.8% (metals basis), turnings
:
Thermo Fisher Scientific
:
7439-95-4
:
:
Mg
:
MFCD00085308
:
24.3 g/mol
:
Ambient
:
231-104-6
:
1869
:
4.1,III
:
GHS02
msds_ALFA010232
Thermo Scientific Acroseal 25 Years