
Magnesium ≥99.6% (metals basis excluding Ca), powder -100+200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-95-4
:
Formula:Mg
MDL Number:MFCD00085308
:24.3 g/mol
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1.74 g/cm³ (25 °C)
Flash Pt:500 °C
Storage Temperature:Ambient
EINECS:231-104-6
UN:1418
ADR:4.3,II
:GHS02