Order Entry
Export
ContactUsLinkComponent
Magnesium, ribbon, Sigma-Aldrich®
Magnesium, ribbon, Sigma-Aldrich®
Catalog # SIAL13103-6X25G
Supplier:  Merck
CAS Number:  
Magnesium, ribbon, Sigma-Aldrich®
Catalog # SIAL13103-6X25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    150 g
  • Seq:
    3475

Specifications

About this item

 :
 :24.3 g/mol
Merck Index:13,05675
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1.74 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-104-6
UN:1869
ADR:4.1,III
 :M-Clarity™ quality level = MQ100
CAS Number:7439-95-4
Formula:Mg
MDL Number:MFCD00085308