Magnesium 99+%, ribbon
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-95-4
:
Formula:Mg
MDL Number:MFCD00085308
:24.3 g/mol
Merck Index:13,05675
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1,74 g/cm³ (20 °C)
Flash Pt:500 °C
Storage Temperature:Ambient
EINECS:231-104-6
UN:1869
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Silver white Metallic ribbon
- Assay:≥99.0 %
- Copper (Cu):≤50 ppm
- Iron (Fe):≤500 ppm
- Lead (Pb):≤50 ppm
- Nickel (Ni):≤50 ppm
- Zinc (Zn):≤200 ppm