

Lithium, Sigma-Aldrich®
Catalog # SIAL278327-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:6.94 g/mol
Merck Index:13,05542
Boiling Pt:1317 °C (1013 hPa)
Melting Pt:179 °C
Density:0.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-102-5
UN:1415
ADR:4.3,I
:GHS02
:GHS05
CAS Number:7439-93-2
Formula:Li
MDL Number:MFCD00134051