Reagent reservoir
Catalog # NEOL7-4751
Supplier: NEOLAB
Specifications
- Capacity:100 ml
- Colour:Opaque
- W×D×H:90×20×20 mm
- Pk:10
Specifications
About this item
This small volume reservoir is made from opaque PP and can be used with single channel or 8-channel pipettes.
- Can be connected with other reservoirs
- Autoclavable