Anti-IgG Rabbit Polyclonal Antibody (Unconjugated)
Catalog # IMMRIR2369
Supplier: ImmunoReagents
Specifications
- Antibody type:Control
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Form:Liquid
- Antibody fragment:F(ab)'2
- Storage temperature:Cool
- Concentration:> 1.0 mg/ml (E 1% at 280 nm = 13.0)
- Shipping temperature:2-8 °C
- Size:2 mg
- Pk:2 mg
Specifications
About this item
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: