Order Entry
Export
ContactUsLinkComponent
Protein kinase C (19-31) (2)
Protein kinase C (19-31) (2)
  ENZOBMLP2060001
 :  
Protein kinase C (19-31) (2)
  ENZOBMLP2060001
 :  

 

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    Protein kinase C (19-31) (2)
  • Purity:
    >97% (HPLC)
  • Formulation:
    TFA salt. Lyophilised.

 

 

PKC inhibitor

Shorter and more potent inhibitor of PKC (IC50=100 nM) than Protein Kinase C (19-36), Pseudosubstrate Inhibitor (BML-P150).