Protein kinase C (19-36)
Catalog # ENZOBMLP1500500
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:0,5 mg
- Seq:02
- Protein/peptide name:Protein kinase C (19-36)
- Purity:97%
Specifications
About this item
PKC inhibitor
Corresponds to PKC pseudosubstrate regulatory sequence. Potent and selective PKC inhibitor which binds to the active site of the kinase. IC50 for purified brain PKC=0,18 µM, PKA=423 µM, MLCK=24 µM.