Order Entry
Export
ContactUsLinkComponent
Z-YVAD(OMe)-FMK
Z-YVAD(OMe)-FMK
Catalog # ENZOALX260074M001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Z-YVAD(OMe)-FMK
Catalog # ENZOALX260074M001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    Z-YVAD(OMe)-FMK
  • Purity:
    >98% (TLC)

Specifications

About this item

Inhibitor of caspase-1 and -4

Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4.