Z-IE(OMe)TD(OMe)-Fmk
ENZOALX260073M001
:
- Pk:1 mg
- Seq:02
- Protein/peptide name:Z-IE(OMe)TD(OMe)-FMK
- Purity:>90% (HPLC)
Caspase and granzyme inhibitor
Irreversible, cell permeable inhibitor of caspase-8 and granzyme B. Inhibits also caspase-10.