Order Entry
Export
ContactUsLinkComponent
Ac-AAVALLPAVLLALLAPYVAD-CHO
Ac-AAVALLPAVLLALLAPYVAD-CHO
Catalog # ENZOALX260047M001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Ac-AAVALLPAVLLALLAPYVAD-CHO
Catalog # ENZOALX260047M001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    Ac-AAVALLPAVLLALLAPYVAD-CHO
  • Purity:
    >97% (HPLC)

Specifications

About this item

Caspase-1 inhibitor

Reversible, cell permeable inhibitor of caspase-1.