Orders LinkContactUsLinkComponent
Ras inhibitory peptide
Ras inhibitory peptide
Supplier:   Enzo Life Sciences
Ras inhibitory peptide
Supplier:  Enzo Life Sciences
Certificates
2 Options Available Below
Product Details & Documents
The peptide sequence corresponds to amino acid residues 1149-1158 of the guanine nucleotide-releasing factor hSos1, which is essential for the control of Ras activity. Blocks the binding of hSos1 to human Grb2, a protein that binds to activated receptor tyrosine kinases.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • ENZOALX166001M001
  • ENZOALX166001M005
  • Protein/peptide name:
  • Ras inhibitory Peptide
  • Ras inhibitory Peptide
  • Purity:
  • >97%
Specifications
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 200,00 €
    200,00 €
    1 * 1 mg
       200,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ENZOALX166001M001
    Protein/peptide name
    Ras inhibitory Peptide
    Purity
    >97%
  • 5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 796,00 €
    796,00 €
    1 * 5 mg
       796,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ENZOALX166001M005
    Protein/peptide name
    Ras inhibitory Peptide
    Purity