Salmonella minnesota R595 (Re) Lipid A
Catalog # ENZOALX581201L002
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:2 mL
- Species:Salmonella minnesota R595 (Re)
- Seq:02
- Enzyme name:Lipid A
- Purity:≥99.9% (regarding protein or DNA contaminants; KDO not detectable)
- Concentration:1 mg/ml
Specifications
About this item
Isolated from Salmonella minnesota R595 (Re mutant). Prepared by mild acid hydrolysis from purified Salmonella minnesota R595 (Re) LPS (isolated by a modification of the PCP extraction method), converted to the uniform salt form and dissolved in sterile pyrogen-free double distilled water.
Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water.