Order Entry
Export
ContactUsLinkComponent
Human aminopeptidase N (from serum)
Human aminopeptidase N (from serum)
Catalog # ENZOBMLSE5420010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human aminopeptidase N (from serum)
Catalog # ENZOBMLSE5420010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Source:
    Serum
  • Species:
    Human
  • Seq:
    02
  • Enzyme name:
    Aminopeptidase N
  • Purity:
    Purified

Specifications

About this item

Isolated from human serum.