Order Entry
Export
ContactUsLinkComponent
Diaphorase
Diaphorase
Catalog # ENZOENZPRT1550500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Diaphorase
Catalog # ENZOENZPRT1550500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 Vial
  • CAS No.:
    9001-68-7
  • Seq:
    02
  • Protein synonyms:
    NADPH dehydrogenase
  • Protein/peptide name:
    Diaphorase
  • Purity:
    >95%
  • Molecular weight:
    ~56 kDa
  • Formulation:
    Liquid. In 50 mm TRIS, pH 8,0, containing 150 mm NaCl
  • Purification:
    Purified by multi-step chromatography

Specifications

About this item

Produced in E. coli.