Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant IL-10

Human recombinant IL-10

10 µG
Catalog # ENZOENZPRT1340010
Supplier:   Enzo Life Sciences
Human recombinant IL-10
Catalog # ENZOENZPRT1340010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant IL-10

10 µG

CAS Number:  
Price
305,00 €
Log in for availability
1 * 10 µG
Product Details & Documents

Produced in E.coli.

Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    ENZOENZPRT1340010
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-10
  • Formulation:
    Lyophilized from 5 mm sodium phosphate, pH 7,2
  • Protein synonyms:
    Cytokine synthesis inhibitory factor|Interleukin-10|CSIF
  • Purity:
    >98% (SDS-PAGE gel & HPLC)
  • Species:
    Human
  • Molecular weight:
    ~18.6 kDa

Specifications