Order Entry
Export
ContactUsLinkComponent
Human Recombinant 14-3-3epsilon (GST-tag) (from E. coli), GST Tag
Human Recombinant 14-3-3epsilon (GST-tag) (from E. coli), GST Tag
Catalog # ENZOBMLSE3140100
Supplier:  Enzo Life Sciences
CAS Number:  
Human Recombinant 14-3-3epsilon (GST-tag) (from E. coli), GST Tag
Catalog # ENZOBMLSE3140100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    GST Tag
  • Protein/peptide name:
    14-3-3 epsilon (GST-tag)

Specifications

About this item

Produced in E. coli. Human 14-3-3epsilon is fused at the N-terminus to a GST-tag.