Order Entry
Export
ContactUsLinkComponent
[beta-Ala8]Neurokinin A (4-10)
[beta-Ala8]Neurokinin A (4-10)
Catalog # ENZOBMLP5060001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
[beta-Ala8]Neurokinin A (4-10)
Catalog # ENZOBMLP5060001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    [beta-Ala8]Neurokinin A (4-10)
  • Purity:
    97%

Specifications

About this item

Neurokinin receptor ligand

Highly selective NK2 neurokinin receptor agonist.