Order Entry
Export
Orders LinkContactUsLinkComponent
WP-AMC

WP-AMC

5 mg
Catalog # ENZOBMLP4500005
Supplier:   Enzo Life Sciences
WP-AMC
Catalog # ENZOBMLP4500005
Supplier:  Enzo Life Sciences
CAS Number:  
CAS Number:  
Price
139,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

DPP substrate

Fluorogenic substrate likely cleaved by dipeptidyl peptidase IV (DPIV, DPPIV, CD26), DPP8, and other prolyl peptidases. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em = 460 nm.

Specifications

Specifications

  • Pk:
    5 mg
  • Cat. No.:
    ENZOBMLP4500005
  • Protein/peptide name:
    WP-AMC
  • Formulation:
    Lyophilised off-white solid
  • Purity:
    >96% (HPLC)

Specifications