Order Entry
Export
ContactUsLinkComponent
YEEI peptide
YEEI peptide
Catalog # ENZOBMLP3000001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
YEEI peptide
Catalog # ENZOBMLP3000001
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    YEEI Peptide
  • Purity:
    >98%

Specifications

About this item

Src kinase activator

Sequence is from hamster polyoma virus middle T inclusive of Tyr324. This 11-residue phosphopeptide activates Src family members by binding with high affinity (Kd=3-6 nM for Src SH2) to their SH2 domains. The crystal structures of this peptide complexed to Src and Lck SH2 domains have been determined.