Compound 48/80
Catalog # ENZOBMLP2270001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Compound 48/80
- Purity:98%
Specifications
About this item
Protease substrate
Fluorogenic substrate for ECE-1 and ECE-2, ACE1 and ACE2, neprilysin, MMP-2 and MMP-9, thimet oligopeptidase, as well as cathepsin A, cathepsin X/Z, BACE, and insulinase, but not MMP-1.Mca fluorescence is quenched by the Dnp group until cleavage separates them. Ex.: 328 nm, Em.: 393 nm, although the following Ex/Em have also been used: 320-340/400-420. This substrate is useful for inhibitor screening and kinetic analysis.