OmniMMP™ fluorogenic Substrate
Catalog # ENZOBMLP1260001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:OmniMMP™ fluorogenic Substrate
- Purity:98%
Specifications
About this item
Highly quenched and sensitive fluorogenic MMP substrate
Fluorogenic substrate for nearly all matrix metalloproteinase (MMP) enzymes, and ADAM17/TACE. This highly-quenched (99,5%) substrate is useful for inhibitor screening and kinetic analysis. Ex.: 328 nm, Em.: 393 nm, although the following Ex/Em have been used: 320-340/400-420. Also available: OmniMMPTM Fluorogenic Control Peptide (BML-P127).