Order Entry
Export
ContactUsLinkComponent
Mas 17
Mas 17
Catalog # ENZOBMLG4210001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Mas 17
Catalog # ENZOBMLG4210001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    Mas 17
  • Purity:
    98%

Specifications

About this item

Inactive mastoparan analog.

Inactive analog of mastoparan (BML-G410). May be used as a negative control.