Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Mas 7
Mas 7
Mas 7
Catalog # ENZOBMLG4200001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Mas 7
Catalog # ENZOBMLG4200001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
02
Protein/peptide name:
Mas 7
Purity:
>99%
Specifications
About this item
Product Details
Phospholipase activator
Product Details