Order Entry
Export
ContactUsLinkComponent
Microcystis aeruginosa Microcystin HTYR
Microcystis aeruginosa Microcystin HTYR
Supplier:  Enzo Life Sciences
Certificates
undefined
Microcystis aeruginosa Microcystin HTYR
Supplier:  Enzo Life Sciences

Specifications

  • Species:
    Microcystis aeruginosa
  • Protein/peptide name:
    Microcystin HTYR
  • Purity:
    >95% (HPLC)

Specifications