Suc-Leu-Tyr-AMC
Catalog # ENZOALX260054M005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:02
- Protein/peptide name:Suc-Leu-Tyr-AMC
- Purity:>95%
Specifications
About this item
Protease substrate
Substrate for porcine calpain I and II. Also cleaved by papain and E. coli protease Ti. Ex.: 340 to 360 nm, Em.: 440 to 460 nm. This substrate is useful for inhibitor screening and kinetic analysis.