Ac-IETD-pNA
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Caspase and granzyme substrate
Chromogenic substrate for caspase-6, -8 and granzyme B. Also a substrate for caspase-10. Similar to Ac-IETD-AMC and Ac-IETD-AFC but cleavage is monitored colorimetrically by absorbance at 405 nm.
Specifications
- Source:
- Species:
- Environmentally Preferable: