Melittin
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
PLA2 activator
Causes disruption of normal cellular activity and cell lysis. Binds calmodulin in a calcium-dependent manner. Activates phospholipase A2 (PLA2) and inhibits protein kinase C (PKC) by binding to the catalytic domain in a Mg-ATP sensitive manner. Antirheumatic.
Specifications
- Protein/peptide name:Melittin
- Purity:>85% (HPLC)