Order Entry
Export
ContactUsLinkComponent
Urinary prostacyclin ELISA kit
Urinary prostacyclin ELISA kit
Catalog # ENZOADI901025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Urinary prostacyclin ELISA kit
Catalog # ENZOADI901025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Description:
    Urinary prostacyclin ELISA kit
  • Seq:
    02
  • Sensitivity:
    6,58 pg/ml (range 7,81 - 2,000 pg/ml)

Specifications

About this item

For the quantitative determination of urinary prostacyclin metabolites in culture supernatants and urine from any species.

The Urinary Prostacyclin ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 by measuring the stable metabolites in one simple assay.

Ordering information: Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-084 (1x96 wells).