Order Entry
Export
Orders LinkContactUsLinkComponent
 

Aldosterone ELISA kit

Supplier:   Enzo Life Sciences
Aldosterone ELISA kit
Supplier:  Enzo Life Sciences
Certificates
Aldosterone ELISA kit

2 Options Available Below

Product Details & Documents

The Aldosterone EIA kit is a colorimetric competitive enzyme immunoassay kit for the quantitative determination of aldosterone in plasma, serum, and urine from any species.

  • Highly sensitive
  • Negligible reactivity with other steroids
  • Fully quantitative results that surpass semi-quantitative Western blots
  • Results overnight + 1 hour

 : Kit includes DxS microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ENZOADI900173
  • ENZOADI901173
  • Target protein:
  • Aldosterone
  • Aldosterone
  • Primary antibody reactivity:
  • Species independent
  • Species independent
  • Assay Type:
  • Competitive
  • Competitive
  • Conjugate ELISA:
  • Alkaline Phosphatase (AP)
  • Alkaline Phosphatase (AP)
  • Format:
  • Pre-coated
  • Pre-coated
  • Sensitivity:
  • 4,7 pg/ml (range 3,9 - 250 pg/ml)
  • 4,7 pg/ml (range 3,9 - 250 pg/ml)
  • Sample type:
  • Plasma, serum, and urine samples
  • Plasma, serum, and urine samples
  • Storage conditions:
  • –20 °C (long term)
  • –20 °C (long term)

Specifications

Product Family Options

Product Information

  • Tests per kit
    Pk
    Availability
    Price
  • ENZOADI900173
    Aldosterone ELISA kit
    Aldosterone ELISA kit
    1×96 wells
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 468,00 €
    468,00 €
    1 * 1 KIT
       468,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ENZOADI900173
    Target protein
    Aldosterone
    Primary antibody reactivity
    Species independent
    Assay Type
    Competitive
    Conjugate ELISA
    Alkaline Phosphatase (AP)
    Format
    Pre-coated
    Sensitivity
    4,7 pg/ml (range 3,9 - 250 pg/ml)
    Sample type
    Plasma, serum, and urine samples
    Storage conditions
    –20 °C (long term)
  • ENZOADI901173
    Aldosterone ELISA kit
    Aldosterone ELISA kit
    5×96 wells
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 880,00 €
    1 880,00 €
    1 * 1 KIT
       1 880,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ENZOADI901173
    Target protein
    Aldosterone
    Primary antibody reactivity
    Species independent
    Assay Type
    Competitive
    Conjugate ELISA
    Alkaline Phosphatase (AP)
    Format
    Pre-coated
    Sensitivity
    4,7 pg/ml (range 3,9 - 250 pg/ml)
    Sample type
    Plasma, serum, and urine samples
    Storage conditions
    –20 °C (long term)