Order Entry
Export
ContactUsLinkComponent
[pSer326]HSF1 ELISA Kits
[pSer326]HSF1 ELISA Kits
Catalog # ENZOADI900199
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
[pSer326]HSF1 ELISA Kits
Catalog # ENZOADI900199
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    [pSer326]HSF1
  • Description:
    [pSer326]HSF1 ELISA kit
  • Seq:
    02
  • Storage conditions:
    –20 °C (long term and short term)
  • Sensitivity:
    61 pg/ml

Specifications

About this item

The HSF1 EIA kit is a complete kit for the quantitative determination of HSF1 phosphorylated at Ser326 in cell lysates and tissue extracts from human and mouse origins. This is a sensitive ELISA kit for the key transcription factor for heat shock protein (chaperone) research.

  • Use less precious sample compared to Western blot
  • Time saving and convenient
  • Ready to use liquid color coded reagents and pre-coated plate

For the quantitative determination of human and mouse [pSer326]HSF1 in cell lysates and tissue.

Delivery: Kit includes antibody, assay buffer, conjugate, extraction reagent #1, microtiter plate, standard, stop solution, TMB substrate, wash buffer concentrate.