Order Entry
Export
ContactUsLinkComponent
 
Anti-CRYAA Rabbit Polyclonal Antibody
 
Anti-CRYAA Rabbit Polyclonal Antibody

 

  • Catalog No:
  • ENZOADISPA224F
  • ENZOADISPA224D
  • Antigen symbol:
  • CRYAA
  • CRYAA
  • Antigen name:
  • Crystallin Alpha A
  • Crystallin Alpha A
  • Antigen synonyms:
  • HSPB4|CTRCT9|CRYA1
  • HSPB4|CTRCT9|CRYA1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human, Bovine, Mouse
  • Human, Bovine, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS, pH 7,2, containing 0,09% sodium azide and 50% glycerol.
  • Liquid. In PBS, pH 7,2, containing 0,09% sodium azide and 50% glycerol.
  • Purification:
  • Protein A affinity purified
  • Protein A affinity purified