Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: 9G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Pig, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine
Specifications
- Catalog No:
- ENZOADISPA850PEF
- ENZOADISPA850PED
- Antigen symbol:
- HSP90B1
- HSP90B1
- Antigen name:
- Heat shock protein 90kDa beta member 1
- Heat shock protein 90kDa beta member 1
- Antigen synonyms:
- tumor rejection antigen 1|glucose regulated protein|ECGP|endoplasmin|GP96|GRP-94|94 kDa|heat shock protein 90 kDa beta member 1|Tumor rejection antigen-1|94 kDa glucose-regulated protein|GRP94|TRA1|endothelial cell (HBMEC) glycoprotein|gp96 homolog
- tumor rejection antigen 1|glucose regulated protein|ECGP|endoplasmin|GP96|GRP-94|94 kDa|heat shock protein 90 kDa beta member 1|Tumor rejection antigen-1|94 kDa glucose-regulated protein|GRP94|TRA1|endothelial cell (HBMEC) glycoprotein|gp96 homolog
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- PE (Phycoerythrin)
- Clone:
- 9G10
- 9G10
- Reactivity:
- Rat, Mouse, Hamster, Pig, Rabbit, Monkey, Xenopus, Sheep, Chicken, Human, Bovine, Guinea pig, Dog
- Rat, Mouse, Hamster, Pig, Rabbit, Monkey, Xenopus, Sheep, Chicken, Human, Bovine, Guinea pig, Dog
- Host:
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Native chicken Grp94
- Native chicken Grp94
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein G affinity chromatography
- Purified by Protein G affinity chromatography
- Storage buffer:
- PBS, pH 7,2, containing 0,09% sodium azide
- PBS, pH 7,2, containing 0,09% sodium azide
- Storage temperature:
- 4 °C
- 4 °C
- Shipping temperature:
- Blue ice not frozen
- Blue ice not frozen