Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: pan
Specifications
- Catalog No:
- ENZOADISAB6000200
- ENZOADISAB6000050
- Antigen symbol:
- IgG1
- IgG1
- Antigen name:
- Immunoglobulin G1
- Immunoglobulin G1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- MOPC-21
- MOPC-21
- Reactivity:
- Multiple
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein G affinity chromatography
- Purified by Protein G affinity chromatography
- Storage buffer:
- PBS, pH 7,2
- PBS, pH 7,2
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Blue ice not frozen
- Blue ice not frozen