Anti-PKG Rabbit Polyclonal Antibody
Cyclic GMP-dependent protein kinases (PKGs or cGKs) are classified into two types, PKGI and PKGII. Studies have shown that PKGs are highly homologous to PKAs; phosphorylation of cellular proteins by both families of kinases leads to alterations in calcium mobilization, protein phosphatase activity, ion channel function, gene transcription, smooth muscle contractility, and platelet aggregation.
Type: Primary
Antigen: PKG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Pig
- Catalog No:
- ENZOADIKAPPK005F
- ENZOADIKAPPK005D
- Antigen symbol:
- PKG
- PKG
- Antigen name:
- Protein Kinase, Cgmp-Dependent, Type I
- Protein Kinase, Cgmp-Dependent, Type I
- Antigen synonyms:
- cGKI|cGK 1|AAT8|cGKI-BETA|PRKGR1B|cGK|cGKI-alpha|cGK1|PRKG1B|PKG
- cGKI|cGK 1|AAT8|cGKI-BETA|PRKGR1B|cGK|cGKI-alpha|cGK1|PRKG1B|PKG
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Pig, Human, Rat, Mouse
- Pig, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide corresponding to a portion of human PKG
- Synthetic peptide corresponding to a portion of human PKG
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein A affinity chromatography
- Storage buffer:
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice